missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium hydride, 98% (metals basis excluding Mg), Mg <1%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
Description
Solvent in Sol-Gel precursor applications. Calcium hydride is used as an efficient drying agent for aprotic base-stable solvents like ethers and tertiary amines. It is a useful dehydrating agent in a synthesis of aldehyde enamines in high yield and purity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 600°C (decomposition) |
Color | Gray |
Physical Form | Powder |
Quantity | 250 g |
Assay Percent Range | 98% |
Solubility Information | Soluble in water and alcohol. Insoluble in benzene. |
Formula Weight | 42.1 |
Sensitivity | Moisture sensitive |
Appearance | Light gray |
Density | 1.9 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'