missing translation for 'onlineSavingsMsg'
Learn More

Calcium granules, redistilled, -16 mesh, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11360168
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11360168

Brand: Thermo Scientific Alfa Aesar 042917.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-70-2
Molecular Formula Ca
Molecular Weight (g/mol) 40.08
MDL Number MFCD00010897 MFCD00085314
InChI Key OYPRJOBELJOOCE-UHFFFAOYSA-N
Synonym unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate
PubChem CID 5460341
ChEBI CHEBI:29320
IUPAC Name calcium
SMILES [Ca]

Specifications

Appearance Silvery-white granules
Quantity 250 g
Mesh Size -16 Mesh
Odor Odorless
Assay Percent Range 99.5% (Metals basis)
Form Granular
Chemical Name or Material Calcium, Redistilled

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.