missing translation for 'onlineSavingsMsg'
Learn More

Calcium formate, 98%

Product Code. p-7024373
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11403168

Brand: Thermo Scientific Alfa Aesar A18916.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 544-17-2 | C2H2CaO4 | 130.112 g/mol

An industrial chemical compound and faster setting, early strength gain accelerating admixture for concrete or mortars. It is used in the building industry, in the production of feedstuff mixtures, for silage and in the tannery. Also used in leather tanning, producing formic acid, concrete cure accelerator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An industrial chemical compound and faster setting, early strength gain accelerating admixture for concrete or mortars. It is used in the building industry, in the production of feedstuff mixtures, for silage and in the tannery. Also used in leather tanning, producing formic acid, concrete cure accelerator.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-17-2
Molecular Formula C2H2CaO4
Molecular Weight (g/mol) 130.112
MDL Number MFCD00036108
InChI Key CBOCVOKPQGJKKJ-UHFFFAOYSA-L
Synonym calcium formate, calcium diformate, formic acid, calcium salt, calcoform, unii-np3jd65npy, mravencan vapenaty czech, calcium formate ca hco2 2, formic acid calcium salt, np3jd65npy, formic acid, calcium salt 2:1
PubChem CID 10997
ChEBI CHEBI:81851
IUPAC Name calcium;diformate
SMILES C(=O)[O-].C(=O)[O-].[Ca+2]

Specifications

Melting Point >300°C
Density 2.02
Quantity 100 g
Solubility Information Soluble in water.
Formula Weight 130.12
Percent Purity 98%
Chemical Name or Material Calcium formate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.