missing translation for 'onlineSavingsMsg'
Learn More

Calcium fluoride sputtering target, 12.7mm (0.5 in.) dia. x 2.5mm (0.098 in.) thick, 99.95% (metals basis), Thermo Scientific™

Catalog Number 15494895
Click to view available options
Quantity:
1 Ea.
This item is not returnable. View return policy
This item is not returnable. View return policy

Magnetron sputtering source

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS 7789-75-5
Quantity 1 Ea.
Molecular Formula CaF2
MDL Number MFCD00010907
Synonym calcium fluoride, kalziumfluorid, kalziumdifluorid, calcium 2+ difluoride, acmc-1bk0c, ksc377c5h, calcium fluoride 100g, calcium fluoride,optical grade
InChI Key WUKWITHWXAAZEY-UHFFFAOYSA-L
SMILES [F-].[F-].[Ca++]
IUPAC Name calcium;difluoride
Molecular Weight (g/mol) 78.08
PubChem CID 84512
ChEBI CHEBI:35437
Formula Weight 78.08
Percent Purity 99.95%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Calcium fluoride sputtering target, 12.7mm (0.5 in.) dia. x 2.5mm (0.098 in.) thick
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'