missing translation for 'onlineSavingsMsg'
Learn More

Calcium fluoride, Puriss., meets analytical spec. of DAB, 99.0-102.0%, Solstice

Product Code. 15640540 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
1 kg
40 kg
Unit Size:
100g
1kg
40kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15640540 40 kg 40kg
15620540 100 g 100g
15630540 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15640540 Supplier Honeywell-Fluka Supplier No. 0112340KG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss., meets analytical specification of DAB, 99.0-102.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-75-5
Molecular Formula CaF2
Molecular Weight (g/mol) 78.08
MDL Number MFCD00010907
InChI Key WUKWITHWXAAZEY-UHFFFAOYSA-L
Synonym calcium fluoride, kalziumfluorid, kalziumdifluorid, calcium 2+ difluoride, acmc-1bk0c, ksc377c5h, calcium fluoride 100g, calcium fluoride,optical grade
PubChem CID 84512
ChEBI CHEBI:35437
IUPAC Name calcium;difluoride
SMILES [F-].[F-].[Ca++]

Specifications

CAS Max % 100.0000%
Quantity 40 kg
Assay Percent Range 99.0 to 102.0%
UN Number NONH for all modes of transport
Formula Weight 78.07
Percent Purity 99.0 to 102.0%
Grade puriss.
Loss on Drying max. 0.5%
Packaging Fiberboard Box
Density 3.18g/mL at 25°C
Chemical Name or Material Calcium fluoride, Puriss., meets analytical specification of DAB, 99.0-102.0%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.