missing translation for 'onlineSavingsMsg'
Learn More

Calcium dihydrogen phosphate hydrate, 97%

Product Code. 11490063
Change view
Click to view available options
Quantity:
10,000 g
2500 g
500 g
Unit Size:
10000g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11490063 2500 g 2500g
11480063 500 g 500g
11400073 10,000 g 10000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490063 Supplier Thermo Scientific Alfa Aesar Supplier No. A12123.0E
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 301524-28-7 | CaH4O8P2 | 234.05 g/mol

Calcium dihydrogen phosphate hydrate is a pH control reagent, stabilizer for plastics, manufacture of glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Calcium dihydrogen phosphate hydrate is a pH control reagent, stabilizer for plastics, manufacture of glass.

Solubility
Soluble in water 18 g/L @ 30°C.

Notes
Loses H{2}O at 100°, dec. at 200°
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 301524-28-7
Molecular Formula CaH4O8P2
Molecular Weight (g/mol) 234.05
MDL Number MFCD00010898
InChI Key YYRMJZQKEFZXMX-UHFFFAOYSA-L
Synonym Calcium phosphate, monobasic; Calcium tetrahydrogen diphosphate
IUPAC Name calcium didihydrogen phosphate
SMILES [Ca++].OP(O)([O-])=O.OP(O)([O-])=O

Specifications

Melting Point ∼200°C (decomposition)
Quantity 2500 g
Flash Point None
Merck Index 14,1693
Solubility Information Soluble in water 18g/L @ 30°C.
Formula Weight 234.06 (Anhydrous)
Percent Purity 97%
Chemical Name or Material Calcium dihydrogen phosphate hydrate
compliance-icons
Product Identifier
  • Calcium dihydrogen phosphate hydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Calcium dihydrogen phosphate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.