missing translation for 'onlineSavingsMsg'
Learn More

Calcium D-gluconate monohydrate, TRC

Product Code. 30134033
Change view
Click to view available options
Quantity:
100 mg
5 g
100 g
Unit Size:
100g
100mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134033 5 g 5g
30011103 100 g 100g
30009351 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134033 Supplier Toronto Research Chemicals Supplier No. TRCC1451035G
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66905-23-5
Molecular Formula 2 C6 H11 O7 . Ca . H2 O
Molecular Weight (g/mol) 448.39
Synonym Calcium bis[(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate] monohydrate, Calcium di(D-gluconate) monohydrate, Calcium Gluconate Monohydrate
IUPAC Name calcium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate;hydrate
SMILES O.[Ca+2].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(=O)[O-].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(=O)[O-]

Specifications

Quantity 5 g
InChI Formula InChI=1S/2C6H12O7.Ca.H2O/c2*7-1-2(8)3(9)4(10)5(11)6(12)13;;/h2*2-5,7-11H,1H2,(H,12,13);;1H2/q;;+2;/p-2/t2*2-,3-,4+,5-;;/m11../s1
Formula Weight 448.0741
Chemical Name or Material Calcium Gluconate Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.