missing translation for 'onlineSavingsMsg'
Learn More

Calcium D-gluconate gel, 2.5% w/w aq. soln., Thermo Scientific Chemicals

Catalog Number p-7059409
Click to view available options
Quantity:
30 g
60 g
299-28-5
C12H22CaO14
430.372
MFCD00064209
NEEHYRZPVYRGPP-IYEMJOQQSA-L
calcium gluconate, calcium d-gluconate, calciofon, calglucon, glucobiogen, ebucin, calcicol, calcipur, calglucol, dragocal
9290
calcium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
C(C(C(C(C(C(=O)[O-])O)O)O)O)O.C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Ca+2]
299-28-5
C12H22CaO14
430.372
MFCD00064209
NEEHYRZPVYRGPP-IYEMJOQQSA-L
calcium gluconate, calcium d-gluconate, calciofon, calglucon, glucobiogen, ebucin, calcicol, calcipur, calglucol, dragocal
9290
calcium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
C(C(C(C(C(C(=O)[O-])O)O)O)O)O.C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Ca+2]

Calcium D-gluconate gel is used to treat hydrofluoric acid burns. It is used as a cardioprotective agent in hyperkalemia, antidote for black widow spider envenomation and an antidote for magnesium sulfate toxicity. It is also involved in the treatment of hypocalcemia. Further, it is used as skin conditioning as well as provides the source of calcium ions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 30 g
Solubility Information Slowly soluble in water. Insoluble in alcohol,acetic acid and organic solvents.
Formula Weight 430.37
Concentration or Composition (by Analyte or Components) 2.5% w/w aq. soln.
Physical Form Gel
Chemical Name or Material Calcium D-gluconate gel

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'