missing translation for 'onlineSavingsMsg'
Learn More

Calcium cyanamide, tech.

Product Code. 11493780
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11493780 500 g 500g
11473780 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11493780 Supplier Thermo Scientific Alfa Aesar Supplier No. 089462.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 156-62-7 | CCaN2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 156-62-7
Molecular Formula CCaN2
MDL Number MFCD00064894

Specifications

Melting Point 1340°C
Color Gray
Physical Form Powder
Quantity 500 g
Linear Formula CaNCN
UN Number UN1403
Beilstein 4124391
Merck Index 14,1662
Formula Weight 80.11
Grade Technical
Odor Specific type
Sensitivity Moisture sensitive
Density 2.29 g/mL
Chemical Name or Material Calcium cyanamide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.