missing translation for 'onlineSavingsMsg'
Learn More

Calcium citrate tribasic tetrahydrate, 98+%, for analysis

Product Code. 10445025 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10445025 250 g Plastic bottle 250g
10657974 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10445025 Supplier Thermo Scientific Acros Supplier No. 390082500
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 5785-44-4 | C12H10Ca3O14·4H2O | 570.5 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5785-44-4
Molecular Formula C12H10Ca3O14·4H2O
Molecular Weight (g/mol) 570.5
MDL Number MFCD00150786
InChI Key LNIZKKFWMDARJV-UHFFFAOYSA-H
Synonym calcium citrate tetrahydrate, unii-mlm29u2x85, calcium citrate usp, tricalcium dicitrate tetrahydrate, citric acid calcium salt, citracal tn, calcium citrate 3:2 , tetrahydrate, acmc-20aliq, ksc493e4f
PubChem CID 5282392
IUPAC Name tricalcium;2-hydroxypropane-1,2,3-tricarboxylate;tetrahydrate
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.O.O.[Ca+2].[Ca+2].[Ca+2]

Specifications

Loss on Drying 10.0 % to 13.3 % (150°C, 4 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98.0 to 102% (on dried basis) (Complexometry)
Packaging Plastic bottle
Quantity 250 g
Merck Index 15, 1663
Solubility Information Solubility in water: 1g/L (20°C)
Formula Weight 570.5
Percent Purity 98+%
Grade Analysis
Insoluble Matter 0.2% max. (in HCl)
Physical Form Liquid
Chemical Name or Material Calcium citrate tribasic tetrahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.