missing translation for 'onlineSavingsMsg'
Learn More

Calcium Citrate Tetrahydrate, TRC

Product Code. 30330536
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30330536 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30330536 Supplier Toronto Research Chemicals Supplier No. TRCC14507550G
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Calcium Citrate Tetrahydrate
CAS 5785-44-4
Molecular Formula 2 C6 H5 O7 . 3 Ca . 4 H2 O
Quantity 50 g
Synonym 1,2,3-Propanetricarboxylic acid, 2-hydroxy-, calcium salt, hydrate (2:3:4), 1,2,3-Propanetricarboxylic acid, 2-hydroxy-, calcium salt (2:3), tetrahydrate (9CI), Citric acid, calcium salt (2:3), tetrahydrate (8CI), Calcium citrate tetrahydrate, Tricalcium dicitrate tetrahydrate
InChI Formula InChI=1S/2C6H8O7.3Ca.4H2O/c2*7-3(8)1-6(13,5(11)12)2-4(9)10;;;;;;;/h2*13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;;4*1H2/q;;3*+2;;;;/p-6
SMILES O.O.O.O.[Ca+2].[Ca+2].[Ca+2].OC(CC(=O)[O-])(CC(=O)[O-])C(=O)[O-].OC(CC(=O)[O-])(CC(=O)[O-])C(=O)[O-]
IUPAC Name tricalcium;2-hydroxypropane-1,2,3-tricarboxylate;tetrahydrate
Molecular Weight (g/mol) 570.49
Formula Weight 569.9371
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.