missing translation for 'onlineSavingsMsg'
Learn More

Calcium citrate tetrahydrate, 96%

Product Code. 11422587
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11422587 100 g 100g
11432587 500 g 500g
11442587 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11422587 Supplier Thermo Scientific Alfa Aesar Supplier No. A14828.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 5785-44-4 | C12H18Ca3O18 | 570.492 g/mol

Calcium citrate tetrahydrate may be used as raw material in the preparation of nanoparticle with the nominal composition Ca0.8 Ba0.2 Ti03 :Pr3+, via sol-gel process. It may be employed as carbon source in a simple and effective carbonization method to prepare porous carbon without any activation process. It is suitable for use in the preparation of bovine serum albumin (BSA) nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Calcium citrate tetrahydrate may be used as raw material in the preparation of nanoparticle with the nominal composition Ca0.8 Ba0.2 Ti03 :Pr3+, via sol-gel process. It may be employed as carbon source in a simple and effective carbonization method to prepare porous carbon without any activation process. It is suitable for use in the preparation of bovine serum albumin (BSA) nanoparticles.

Solubility
Soluble in water and hydrochloric acid.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5785-44-4
Molecular Formula C12H18Ca3O18
Molecular Weight (g/mol) 570.492
MDL Number MFCD00150786
InChI Key LNIZKKFWMDARJV-UHFFFAOYSA-H
Synonym calcium citrate tetrahydrate, unii-mlm29u2x85, calcium citrate usp, tricalcium dicitrate tetrahydrate, citric acid calcium salt, citracal tn, calcium citrate 3:2 , tetrahydrate, acmc-20aliq, ksc493e4f
PubChem CID 5282392
IUPAC Name tricalcium;2-hydroxypropane-1,2,3-tricarboxylate;tetrahydrate
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.O.O.[Ca+2].[Ca+2].[Ca+2]

Specifications

Quantity 100 g
Solubility Information Soluble in water and hydrochloric acid.
Formula Weight 572.60 (500.54 Anhydrous)
Percent Purity 96%
Chemical Name or Material Calcium citrate tetrahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.