missing translation for 'onlineSavingsMsg'
Learn More

Calcium chromate, 99.9% (metals basis)

Product Code. 11389498
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11389498

Brand: Thermo Scientific Alfa Aesar 043333.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13765-19-0 | CaCrO4 | 156.07 g/mol

Calcium chromate is used as a corrosion inhibitor and pigment. It is also used as a depolarizer in batteries and a metal coating and primer. Further, it is used in electroplating, photochemical processing and industrial waste treatment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13765-19-0
Molecular Formula CaCrO4
Molecular Weight (g/mol) 156.07
MDL Number MFCD00049403
InChI Key RFAFBXGYHBOUMV-UHFFFAOYSA-N
Synonym calcium chromate, gelbin, calcium monochromate, yellow ultramarine, calcium chrome yellow, chromatite syn, calcium chromate vi, sintered calcium chromate, unii-j1fgh4zj4m, chromic acid, calcium salt
PubChem CID 26264
IUPAC Name calcium;dioxido(dioxo)chromium
SMILES [Ca++].[O-][Cr]([O-])(=O)=O

Specifications

Color Yellow
Physical Form Powder
Quantity 100 g
UN Number UN3087
Solubility Information Soluble in acid and water. Insoluble in alcohol.
Formula Weight 156.07
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Calcium chromate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.