Learn More
Calcium chloride hydrate, Puratronic™, 99.9965% (metals basis)
Description
It is used as a food additive, as an electrolyte, it is used as a buffer in pools, an additive in plastics and in fire extinguishers. It is used as a fabric softener, dust control on roads. Calcium chloride is a soluble compound that can serve as a source of calcium ions in a solution. Calcium plays important roles in many biological processes, including signal transduction, muscle contraction, maintenance of cell membranes and cell wall stability.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Crystalline Aggregates |
| Quantity | 50 g |
| Merck Index | 14,1659 |
| Solubility Information | Soluble in water (5360mg/ml at 20°C),and alcohol. |
| Formula Weight | 110.99 (Anhydrous) |
| Percent Purity | 99.9965% |
| Grade | Puratronic |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Density | 1.71 g/mL |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.