missing translation for 'onlineSavingsMsg'
Learn More

Calcium Chloride Hexahydrate, Honeywell Fluka™

Product Code. 15671100 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
6 x 1 kg
6 x 2.5 kg
Unit Size:
1kg
2.5kg
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15671100 1 kg 1kg
15681100 2.5 kg 2.5kg
15691100 6 x 1 kg Pack of 6
15601110 6 x 2.5 kg Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15671100 Supplier Honeywell-Fluka Supplier No. 120741KG
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

meets analytical specification of European Pharmacopoeia, E509, 97-103%, ≤0.0001% Al

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7774-34-7
Molecular Formula CaCl2H12O6
Molecular Weight (g/mol) 219.068
MDL Number MFCD00149616
InChI Key QHFQAJHNDKBRBO-UHFFFAOYSA-L
Synonym calcium chloride hexahydrate, antarcticite, acmc-20alis, calcium hexahydrate dichloride, calcium chloride-water 1/6, calcium chloride hydrate, puratronic, calcium chloride, hydrated, calcium chloride hydrate trace metals basis 1g
PubChem CID 6093252
IUPAC Name calcium;dichloride;hexahydrate
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Ca+2]

Specifications

Quantity 1 kg
UN Number NONH for all modes of transport
Formula Weight 219.08g/mol
Percent Purity 97 to 103%
Density 1.71g/mL at 25°C
Chemical Name or Material Calcium Chloride Hexahydrate, Meets analytical specification of Ph. Eur.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.