missing translation for 'onlineSavingsMsg'
Learn More

Calcium chloride dihydrate, Puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥99%, Solstice

Product Code. 15673380 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
50 kg
6 x 2.5 kg
Unit Size:
1kg
2.5kg
500g
50kg
Pack of 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15673380 2.5 kg 2.5kg
15683380 500 g 500g
15663380 1 kg 1kg
15603390 6 x 2.5 kg Pack of 6
15693380 50 kg 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15673380 Supplier Honeywell-Fluka Supplier No. 313072.5KG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥99%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10035-04-8
Molecular Formula CaCl2 · 2H2O

Specifications

Quantity 2.5 kg
MDL Number MFCD00149613
UN Number NONH for all modes of transport
Formula Weight 147.01g/mol
Percent Purity ≥99%
Grade ACS/puriss. p.a.
Chemical Name or Material Calcium Chloride Dihydrate, Reag. Ph. Eur.
compliance-icons
Product Identifier
  • Calcium chloride dihydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.