missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Chloride, Dihydrate, Granular, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
233.70 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Specifications
Physical Form | Granules |
Quantity | 500 g |
Formula Weight | 147.01 |
Grade | BAKER ANALYZED™ ACS Reagent |
Packaging | Poly Bottle |
Specific Gravity | 1L = 0.84kg |
Chemical Name or Material | Calcium Chloride, Dihydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'