missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Chloride Dihydrate, ≥99.0%, ReagentPlus™ Grade, Honeywell™
Reagent Grade,≥99.0%
£42.59 - £670.00
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15687750
|
Honeywell Research Chemicals
C3881-500G |
500 g | Poly bottle | ||||||
15667750
|
Honeywell Research Chemicals
C3881-1KG |
1 kg | Poly bottle | ||||||
15677750
|
Honeywell Research Chemicals
C3881-2.5KG |
2.5 kg | Poly bottle | ||||||
15697750
|
Honeywell Research Chemicals
C3881-5KG |
5 kg | Drum with liner | ||||||
15657750
|
Honeywell Research Chemicals
C3881-12KG |
12 kg | Fiberboard box | ||||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
10035-04-8 | |
MFCD00149613 |
Specifications
1.90°C (1.013hPa) | |
10035-04-8 | |
4,5 - 8 | |
MFCD00149613 | |
147.01g/mol | |
Odorless |
765°C | |
Colorless | |
Solid | |
NONH for all modes of transport | |
≥99.0% | |
Calcium chloride dihydrate |
Safety and Handling
P305 + P351 + P338
missing translation for 'rtecsNumber' : 233-140-8
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Calcium chloride dihydrate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : calcium chloride-2-hydrate