missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Chloride, Dessicant, ACS Reagent, ≥96.0%, Honeywell™ Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
500g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Boiling Point | ∼1900°C |
| Melting Point | 765°C |
| Color | Colorless |
| pH | ∼6 (at 20°C) |
| Quantity | 100 g |
| Linear Formula | CaCl2 |
| UN Number | NONH for all modes of transport |
| Solubility Information | Completely soluble in water |
| IUPAC Name | calcium dichloride |
| Formula Weight | 110.98g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction