missing translation for 'onlineSavingsMsg'
Learn More

Calcium chloride, Anhydrous, powder, ≥97%, Honeywell Fluka™

Product Code. 15647760 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
5 kg
Packaging:
Poly Bottle
Unit Size:
100g
1kg
2.5kg
500g
5kg
This item is not returnable. View return policy

Product Code. 15647760

Brand: Honeywell-Fluka C49012.5KG

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Anhydrous, powder,≥97%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-52-4
Molecular Formula CaCl2
Molecular Weight (g/mol) 110.98
MDL Number MFCD00010903
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
Synonym calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
PubChem CID 5284359
ChEBI CHEBI:3312
SMILES [Cl-].[Cl-].[Ca++]

Specifications

Boiling Point 1,900°C
Melting Point 772°C (literature)
CAS Max % 100.0000%
Physical Form Powder
Quantity 2.5 kg
Linear Formula CaCl2
UN Number NONH for all modes of transport
IUPAC Name calcium;dichloride
Formula Weight 110.98
Percent Purity >97%
Packaging Poly Bottle
Vapor Pressure 0.01 mmHg (20°C)
Density 2.150 g/cm3
Chemical Name or Material Calcium chloride, Anhydrous, powder, ≥97%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.