missing translation for 'onlineSavingsMsg'
Learn More

Calcium chloride, 2.75% w/v aq. soln., Thermo Scientific Chemicals

Product Code. 11475040
Click to view available options
Quantity:
500 mL
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10035-04-8
MDL Number MFCD00010903

Specifications

Concentration 2.75% w/v aq. soln.
Formula Weight 110.99
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Calcium chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'