missing translation for 'onlineSavingsMsg'
Learn More

Calcium carbonate, ACS, low in alkalies, 99.0% min

Product Code. 11373418
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11373418 1 kg 1kg
11383418 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11373418 Supplier Thermo Scientific Alfa Aesar Supplier No. 036337.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 471-34-1 | CCaO3 | 100.09 g/mol

It is used medicinally as a calcium supplement or as an antacid. It is also used for industrial purposes. Calcium carbonate is the most widely used mineral in the paper, plastics, paints and coatings industries. Hierarchical hollow calcium carbonate particles shows an useful application as anticancer drug carrier. It plays an essential role to catalyze alcoholysis of fats and oils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
Synonym calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
PubChem CID 10112
ChEBI CHEBI:3311
IUPAC Name calcium carbonate
SMILES [Ca++].[O-]C([O-])=O

Specifications

Melting Point 800°C (decomposition)
Color White
Physical Form Powder
Quantity 1 kg
Assay Percent Range 99.0% min.
Solubility Information Soluble in acids,insoluble in water.
Formula Weight 100.09
Grade ACS
Refractive Index 1.6583
Density 2.93
Chemical Name or Material Calcium carbonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.