missing translation for 'onlineSavingsMsg'
Learn More

Calcium carbonate, 98+%, pure, heavy powder, ACROS Organics™

Brand:  Acros Organics 124670010

Code : 29

Additional Details : CAS Number : 471-34-1 Weight : 1.03306kg

Packaging Plastic bottle
Quantity 1kg

 View more versions of this product

Product Code. 10021250

  • £21.79 / 1kg

In stock, ready to ship: 23

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

471-34-1
100.09
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
CHEBI:3311
C(=O)([O-])[O-].[Ca+2]
CCaO3
VTYYLEPIZMXCLO-UHFFFAOYSA-L
10112
calcium;carbonate

Specifications

Calcium carbonate
98% min. (Complexometry)
CaCO3
15,1659
Solubility in water: insoluble. Other solubilities: soluble in diluted acids
C(=O)([O-])[O-].[Ca+2]
100.09
CHEBI:3311
Powder
Pure
Solubility in water: insoluble. Other solubilities: soluble in diluted acids
825.0°C
471-34-1
CCaO3
01,103; 02,57; 04,67; 05,89
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
VTYYLEPIZMXCLO-UHFFFAOYSA-L
calcium;carbonate
10112
100.09
98+%
Plastic bottle
White
1kg
Videos
SDS Documents