missing translation for 'onlineSavingsMsg'
Learn More

Calcium carbonate, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, E170, precipitated, 98.5-100.5% (based on anhydrous substance), Honeywell Fluka™
BIOPHARMA

Product Code. p-7071773 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
25 kg
6 x 1 kg
Unit Size:
1kg
2.5kg
25kg
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15671080 1 kg 1kg
15681080 2.5 kg 2.5kg
15601090 6 x 1 kg Pack of 6
15691080 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15671080 Supplier Honeywell-Fluka Supplier No. 120101KG
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, E170, precipitated, 98.5-100.5% (based on anhydrous substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
Synonym calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
PubChem CID 10112
ChEBI CHEBI:3311
SMILES [Ca++].[O-]C([O-])=O

Specifications

Quantity 1 kg
Linear Formula CaCO3
UN Number NONH for all modes of transport
Beilstein 8008338
IUPAC Name calcium;carbonate
Formula Weight 100.09g/mol
Percent Purity 98.5 to 100.5% (based on anhydrous substance)
Grade puriss.
Density 2.93g/mL at 25°C
Chemical Name or Material Calcium Carbonate(Caco3), Meets analytical specification of Ph. Eur.BP, USP, FCC
Product Identifier
  • Calcium carbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.