missing translation for 'onlineSavingsMsg'
Få mere at vide

Calcium carbonate, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, E170, precipitated, 98.5-100.5% (based on anhydrous substance), Honeywell Fluka™
BIOPHARMA

Artikelnummer. 15691080 Shop alle Honeywell Fluka produkter
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
1 kg
2.5 kg
25 kg
6 x 1 kg
Pakningsstørrelse:
1kg
2.5kg
25kg
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
15691080 25 kg 25kg
15671080 1 kg 1kg
15681080 2.5 kg 2.5kg
15601090 6 x 1 kg Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15691080 Leverandør Honeywell-Fluka Leverandørnr. 1201025KG
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, E170, precipitated, 98.5-100.5% (based on anhydrous substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
Synonym calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
PubChem CID 10112
ChEBI CHEBI:3311
SMILES [Ca++].[O-]C([O-])=O

Tekniske data

CAS Max % 100.0000%
Quantity 25 kg
Linear Formula CaCO3
UN Number NONH for all modes of transport
Beilstein 8008338
IUPAC Name calcium;carbonate
Formula Weight 100.09
Percent Purity ≥98.5%
Grade puriss.
Packaging Paper / Poly Bag
Density 2.93 g/mL at 25 °C(lit.)
Chemical Name or Material Calcium carbonate, Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, E170, precipitated, 98.5-100.5% (based on anhydrous substance)
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.