Learn More
Calcium carbonate, Puratronic™, 99.997% (metals basis)
Description
Calcium carbonate is used in medicine and agriculture. It is also used in flue gas desulfurization in fossil fuel power stations, production of toothpaste, ceramics/glazing applications, purification of iron from iron ore, and to clean tarnish on silver. It is used in the refining of sugar, as filler in thermosetting resins, as an abrasive, as an extender in paints, and as a formation-bridging filtercake-sealing agent in oil industry. It is used in swimming pools for controlling the pH, to improve the rigidity of polypropylene compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 800°C (decomposition) |
| Color | White |
| Physical Form | Powder |
| Quantity | 100 g |
| Assay Percent Range | 99.997% (Metal basis) |
| Solubility Information | Soluble in acids and insoluble in water. |
| IUPAC Name | calcium;carbonate |
| Formula Weight | 100.09 |
| Refractive Index | 1.6583 |
| Appearance | White powder |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.