missing translation for 'onlineSavingsMsg'
Learn More

Calcium carbonate, 98%

Product Code. p-7048986
Change view
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11381749 500 g 500g
11391749 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11381749 Supplier Thermo Scientific Alfa Aesar Supplier No. 012365.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 471-34-1 | CCaO3 | 100.09 g/mol

Manufacture of paint, rubber, plastics, etc. In food, cosmetics, and pharmaceuticals. As soil conditioner, neutralizer of surface waters, and as an industrial acid neutralizerIt is used medicinally as a calcium supplement or as an antacid. It is also used for industrial purposes. Calcium carbonate is the most widely used mineral in paper, plastics, paints and coatings industries. Hierarchical hollow calcium carbonate particles shows an useful application as anticancer drug carrier. It plays an essential role to catalyze alcoholysis of fats and oils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Manufacture of paint, rubber, plastics, etc
  • In food, cosmetics, and pharmaceuticals
  • As soil conditioner, neutralizer of surface waters, and as an industrial acid neutralizer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
Synonym calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
PubChem CID 10112
ChEBI CHEBI:3311
IUPAC Name calcium carbonate
SMILES [Ca++].[O-]C([O-])=O

Specifications

Melting Point 800°C (decomposition)
Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range 98%
Merck Index 14,1657
Solubility Information Practically insoluble in water; Soluble in diluted acids
Formula Weight 100.09
Percent Purity 98%
Odor Odorless
Refractive Index 1.6583
Density 2.930 g/mL
Chemical Name or Material Calcium carbonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.