missing translation for 'onlineSavingsMsg'
Learn More

Calcium carbonate, 97%, pure, chunks, Thermo Scientific Chemicals

Product Code. 10587162 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2 kg
3 kg
Packaging:
Plastic bottle
Unit Size:
2kg
3kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10587162 2 kg Plastic bottle 2kg
10361151 3 kg Plastic bottle 3kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10587162 Supplier Thermo Scientific Acros Supplier No. 403790020
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
Synonym calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
PubChem CID 10112
ChEBI CHEBI:3311
IUPAC Name calcium;carbonate
SMILES [Ca++].[O-]C([O-])=O

Specifications

Melting Point 825.0°C
Color Beige-Gray to Beige-White
Physical Form Chunks
Quantity 2 kg
Assay Percent Range 96% min. (Complexometry)
Linear Formula CaCO3
Fieser 01,103; 02,57; 04,67; 05,89
Merck Index 15, 1659
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in diluted acids
Formula Weight 100.09
Percent Purity 97%
Grade Pure
Packaging Plastic bottle
Chemical Name or Material Calcium carbonate, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.