missing translation for 'onlineSavingsMsg'
Learn More

Calcium bromide, ultra dry , 99.978% (metals basis)

Product Code. 11373857
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11373857 1 g 1g
11363857 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11373857 Supplier Thermo Scientific Alfa Aesar Supplier No. 035684.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7789-41-5 | Br2Ca | 199.886 g/mol

CaBr2 is mainly used as a dense aqueous solution for drilling fluid. It is also used in neuroses medication, freezing mixtures, food preservatives, photography and fire retardants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-41-5
Molecular Formula Br2Ca
Molecular Weight (g/mol) 199.886
MDL Number MFCD00010902
InChI Key WGEFECGEFUFIQW-UHFFFAOYSA-L
Synonym calcium bromide, calcium dibromide, kalziumbromid, brocal, 87cny2eebh, ksc377c0p, calciumbromide, calcium 2+ dibromide, calcium bromide, anhydrous 10g
PubChem CID 5360316
ChEBI CHEBI:31338
IUPAC Name calcium;dibromide
SMILES [Ca+2].[Br-].[Br-]

Specifications

Boiling Point 806°C to 812°C
Melting Point 730°C
Color White
Physical Form Beads
Quantity 1 g
Linear Formula CaBr2
Merck Index 14,1654
Solubility Information It is soluble in water,alcohol and acetone.
Formula Weight 199.9
Percent Purity 99.978%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Density 3.353 g/mL
Assay (metals basis)
Chemical Name or Material Calcium bromide, Ultra dry
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.