missing translation for 'onlineSavingsMsg'
Learn More

Calcium bromide, anhydrous, 99.5%, Thermo Scientific Chemicals

Catalog Number 11335059
Click to view available options
Quantity:
10 g
50 g
250 g
7789-41-5
Br2Ca
199.886
MFCD00010902
WGEFECGEFUFIQW-UHFFFAOYSA-L
calcium bromide, calcium dibromide, kalziumbromid, brocal, 87cny2eebh, ksc377c0p, calciumbromide, calcium 2+ dibromide, calcium bromide, anhydrous 10g
5360316
CHEBI:31338
calcium;dibromide
[Ca+2].[Br-].[Br-]
7789-41-5
Br2Ca
199.886
MFCD00010902
WGEFECGEFUFIQW-UHFFFAOYSA-L
calcium bromide, calcium dibromide, kalziumbromid, brocal, 87cny2eebh, ksc377c0p, calciumbromide, calcium 2+ dibromide, calcium bromide, anhydrous 10g
5360316
CHEBI:31338
calcium;dibromide
[Ca+2].[Br-].[Br-]

For Photography, Dehydrating Agent, Manufacturing of Mineral Water

Calcium bromide is a dehydrating agent used in photography, food preservatives, wood preservatives, fire retardants and freezing mixtures. It is used as an oxidizer for mercury emissions control. It is widely used for the preparation of drilling fluids, completion fluids and drilling muds formulations. It also finds application in the manufacture of mineral water.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 806°C to 812°C
Melting Point 730°C
Color White
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.5%
Solubility Information Soluble in water,methanol,ethanol and acetone. Insoluble in dioxane,chloroform and ether.
Formula Weight 199.9
Sensitivity Hygroscopic
Appearance White to off-white powder or beads
Density 3.353
Chemical Name or Material Calcium bromide, Anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'