missing translation for 'onlineSavingsMsg'
Learn More

Calcium boride, 95+%, Thermo Scientific Chemicals

Catalog Number 11388868
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy
12007-99-7
B6Ca
104.938
MFCD00148904
AWVIDOFQNLOVRC-UHFFFAOYSA-N
calcium boride, calcium hexaboride, b6.ca, calcium hexaboride,-200 mesh, calcium pentacyclo 3.1.0.0 1 , 3 .0 2 ,?.0?,? hexaborane-1,4-diuide, calcium 2+ pentacyclo 3.1.0.0^ 1,3 .0^ 2,4 .0^ 4,6 hexaborane-1,4-diuide
16212529
B12B3[B-]14B5[B-]23B45.[Ca+2]
This item is not returnable. View return policy
12007-99-7
B6Ca
104.938
MFCD00148904
AWVIDOFQNLOVRC-UHFFFAOYSA-N
calcium boride, calcium hexaboride, b6.ca, calcium hexaboride,-200 mesh, calcium pentacyclo 3.1.0.0 1 , 3 .0 2 ,?.0?,? hexaborane-1,4-diuide, calcium 2+ pentacyclo 3.1.0.0^ 1,3 .0^ 2,4 .0^ 4,6 hexaborane-1,4-diuide
16212529
B12B3[B-]14B5[B-]23B45.[Ca+2]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 2235°C
Physical Form Powder
Quantity 50 g
Solubility Information Insoluble in water. Dissolves slowly in acids
Formula Weight 104.95
Percent Purity ≥95%
Density 2.30 g/mL
Chemical Name or Material Calcium boride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'