missing translation for 'onlineSavingsMsg'
Learn More

Calcium bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals

Product Code. 15492707
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 165324-09-4
Molecular Formula C4CaF12N2O8S4
Molecular Weight (g/mol) 600.349
MDL Number MFCD14156019
InChI Key WFABOCFDABTAPE-UHFFFAOYSA-N
Synonym calcium bis trifluoromethylsulfonyl imide, calcium ii bis trifluoromethanesulfonyl imide, calcium ii triflimide, calcium trifluoromethanesulfonimide, calcium bistrifluoromethanesulfonimide, calcium bis trifluoromethylsulfonyl amide, calcium bis bistriflylimide anion, calcium bis trifluoromethyl sulfonyl amide, bis bis trifluoromethylsulfonyl amino calcium, bis trifluoromethanesulfonyl imide calcium ii salt
PubChem CID 46242197
IUPAC Name calcium;bis(trifluoromethylsulfonyl)azanide
SMILES C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Ca+2]

Specifications

Melting Point >300°C
Linear Formula [(CF3SO2)2N]2Ca
Quantity 1 g
UN Number UN1759
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 600.35
Physical Form Solid
Chemical Name or Material Calcium bis (trifluoromethylsulfonyl)imide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'