missing translation for 'onlineSavingsMsg'
Learn More

Calcium benzoate hydrate, 98%, water <8%

Product Code. 11459493
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459493 2500 g 2500g
11439493 100 g 100g
11449493 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11459493 Supplier Thermo Scientific Alfa Aesar Supplier No. A12358.0E
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

CAS: 2090-05-3 | C19H20O4 | 312.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Calcium benzoate hydrate is used in used in the food industry as a preservative.

Solubility
Soluble in water (2.6 g/100ml at 0°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2090-05-3

Specifications

Molecular Formula C14H10CaO4
MDL Number MFCD09953675
Quantity 2500 g
Synonym calcium benzoate, calcium dibenzoate, unii-3qde968mkd, 3qde968mkd, benzoic acid, calcium salt, benzoic acid calcium, acmc-20alj1, calcium 2+ dibenzoate, calcium benzoate, reagent, e213
benzoic acid
Solubility Information Soluble in water (2.6g/100ml at 0°C).
InChI Key NSQPPSOSXWOZNH-UHFFFAOYSA-L
SMILES [Ca++].[O-]C(=O)C1=CC=CC=C1.[O-]C(=O)C1=CC=CC=C1
IUPAC Name calcium;dibenzoate
Molecular Weight (g/mol) 282.31
PubChem CID 62425
Formula Weight 282.32 (Anhydrous)
Percent Purity 98%
Chemical Name or Material Calcium benzoate hydrate, Water <8%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.