missing translation for 'onlineSavingsMsg'
Learn More

Calcium b-hydroxy-b-methylbutyrate, TRC

Product Code. 30347992
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30347992 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30347992 Supplier Toronto Research Chemicals Supplier No. TRCC145683500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Calcium beta-hydroxy-beta-methylbutyrate
CAS 135236-72-5
Purity Grade Notes HPLC
Molecular Formula 2 C5 H9 O3 . Ca
Quantity 500 mg
Synonym 3-Hydroxy-3-methylbutanoic acid calcium salt (2:1)
InChI Formula InChI=1S/2C5H10O3.Ca/c2*1-5(2,8)3-4(6)7;/h2*8H,3H2,1-2H3,(H,6,7);/q;;+2/p-2
SMILES [Ca+2].CC(C)(O)CC(=O)[O-].CC(C)(O)CC(=O)[O-]
IUPAC Name calcium;3-hydroxy-3-methyl-butanoate
Molecular Weight (g/mol) 274.32
Formula Weight 274.0729
Percent Purity >95
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.