missing translation for 'onlineSavingsMsg'
Learn More

Calcium arsenate, 95%

Product Code. 11313618
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Packing group II
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11313618 250 g Packing group II 250g
11323618 50 g Packing group II 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11313618 Supplier Thermo Scientific Alfa Aesar Supplier No. 039219.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 7778-44-1 | As2Ca3O8 | 398.069 g/mol

Calcium arsenate is also used in weather- resistant wood treatment. It was used mainly in viticulture, in the past. it was mainly used against the boll weevil in cotton cultivation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-44-1
Molecular Formula As2Ca3O8
Molecular Weight (g/mol) 398.069
MDL Number MFCD00049407
InChI Key RMBBSOLAGVEUSI-UHFFFAOYSA-H
Synonym calcium arsenate, pencal, cucumber dust, spra-cal, calcium-o-arsenate, chip-cal granular, tricalcium arsenate, calcium orthoarsenate, kalziumarseniat german, arsenic acid, calcium salt
PubChem CID 24501
IUPAC Name tricalcium;trioxido(oxo)-$l^{5}-arsane
SMILES [O-][As](=O)([O-])[O-].[O-][As](=O)([O-])[O-].[Ca+2].[Ca+2].[Ca+2]

Specifications

Color White
Physical Form Powder
Quantity 250 g
Assay Percent Range 95%
Solubility Information It is soluble in water and acids,insoluble in organic solvents
Formula Weight 398.03
Packaging Packing group II
Density 3.62 g/cm3 at 20°C
Chemical Name or Material Calcium arsenate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.