missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Activated Nucleotidase 1/CANT1 Antibody (OTI7F8), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72378AF405
This item is not returnable.
View return policy
Description
Calcium Activated Nucleotidase 1/CANT1 Monoclonal specifically detects Calcium Activated Nucleotidase 1/CANT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Calcium Activated Nucleotidase 1/CANT1 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CANT1 | |
| Recombinant protein fragment corresponding to amino acids 63-401 of human Calcium Activated Nucleotidase 1/CANT1 (NP_620148) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI7F8 | |
| Western Blot, Immunohistochemistry | |
| Apyrase homolog, calcium activated nucleotidase 1, DBQD, EC 3.6.1.6, Putative MAPK-activating protein PM09, Putative NF-kappa-B-activating protein 107, SCAN1, SCAN-1Ca2+-dependent endoplasmic reticulum nucleoside diphosphatase, SHAPYmicromelic dwarfism with vertebral and metaphyseal abnormalities and advancedcarpotarsal ossification, soluble Ca-activated nucleotidase, isozyme 1, soluble calcium-activated nucleotidase 1, soluble calcium-activated nucleotidase SCAN-1 | |
| Mouse | |
| Affinity Purified | |
| Lipid and Metabolism | |
| 124583.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction