missing translation for 'onlineSavingsMsg'
Learn More

Calcium Activated Nucleotidase 1/CANT1 Antibody, Novus Biologicals™

Product Code. 18423171 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18423171 25 μL 25µL
18259396 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18423171 Supplier Novus Biologicals Supplier No. NBP18708125ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 1 publication

Calcium Activated Nucleotidase 1/CANT1 Polyclonal specifically detects Calcium Activated Nucleotidase 1/CANT1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calcium Activated Nucleotidase 1/CANT1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:200
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias Apyrase homolog, calcium activated nucleotidase 1, DBQD, EC 3.6.1.6, Putative MAPK-activating protein PM09, Putative NF-kappa-B-activating protein 107, SCAN1, SCAN-1Ca2+-dependent endoplasmic reticulum nucleoside diphosphatase, SHAPYmicromelic dwarfism with vertebral and metaphyseal abnormalities and advancedcarpotarsal ossification, soluble Ca-activated nucleotidase, isozyme 1, soluble calcium-activated nucleotidase 1, soluble calcium-activated nucleotidase SCAN-1
Gene Symbols CANT1
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:QEENTWFSYLKKGYLTLSDSGDKVAVEWDKDHGVLESHLAEKGRGMELSDLIVFNGKLYSVDDRTGVVYQIEGSKAVPWVILSDGDGTVEKGFKAEWLAVK
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 124583
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.