missing translation for 'onlineSavingsMsg'
Learn More

Calcium Activated Nucleotidase 1/CANT1 Antibody [DyLight 755], Novus Biologicals™

Product Code. 30731385 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30731385 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30731385 Supplier Novus Biologicals Supplier No. AF6720Z

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Calcium Activated Nucleotidase 1/CANT1 Polyclonal antibody specifically detects Calcium Activated Nucleotidase 1/CANT1 in Human samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calcium Activated Nucleotidase 1/CANT1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias Apyrase homolog, calcium activated nucleotidase 1, DBQD, EC 3.6.1.6, Putative MAPK-activating protein PM09, Putative NF-kappa-B-activating protein 107, SCAN1, SCAN-1Ca2+-dependent endoplasmic reticulum nucleoside diphosphatase, SHAPYmicromelic dwarfism with vertebral and metaphyseal abnormalities and advancedcarpotarsal ossification, soluble Ca-activated nucleotidase, isozyme 1, soluble calcium-activated nucleotidase 1, soluble calcium-activated nucleotidase SCAN-1
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human Calcium Activated Nucleotidease 1/CANT1, Gly80-Ile401, Accession # Q8WVQ1
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 124583
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.