missing translation for 'onlineSavingsMsg'
Learn More

Calcium acetate hydrate, Puriss., meets analytical spec. of FCC, E263, 99.0-100.5% (based on anhydrous substance), Solstice
BIOPHARMA

Product Code. 15682710
Change view
Click to view available options
Quantity:
1 kg
25 kg
6 x 1 kg
Unit Size:
1kg
25kg
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15682710 1 kg 1kg
15602720 6 x 1 kg Pack of 6
15692710 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15682710 Supplier Honeywell-Fluka Supplier No. 250111KG
 Restricted Item
Estimated Shipment: 09-02-2027
Add to basket
This item is not returnable. View return policy

Puriss., Meets analytical specification of FCC, E263, 99.0-100.5% (based on anhydrous substance)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114460-21-8
Molecular Formula C4H6CaO4
Molecular Weight (g/mol) 158.17
MDL Number MFCD00012448
InChI Key VSGNNIFQASZAOI-UHFFFAOYSA-L
IUPAC Name calcium diacetate
SMILES [Ca++].CC([O-])=O.CC([O-])=O

Specifications

Quantity 1 kg
Linear Formula (CH3COO)2Ca · xH2O
UN Number NONH for all modes of transport
Beilstein 3692527
Formula Weight 158.17g/mol (anhydrous basis)
Percent Purity 99 to 100.5% (based on anhydrous substance)
Grade puriss.
Chemical Name or Material Calcium Acetate Hydrate, Meets analytical specification of FCC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.