Learn More
Calcium acetate hydrate, Puratronic™, 99.9965% (metals basis)
Description
Calcium acetate has been utilized in the crystallization of proteins, such as human annexin II and the bacterial heme oxygenase (Hmu O), and of a complex of human CDK6 with a virus-encoded cyclin. It has been used in the manufactureof metallic soaps and synthetic resins, in lubricants as a corrosion inhibitor, and as a mordant in dyeing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | (decomposition) |
| Physical Form | Powder |
| Quantity | 5 g |
| Beilstein | 3692527 |
| Merck Index | 14,1646 |
| Solubility Information | Soluble in water. Slightly soluble in methanol,hydrazine insoluble in acetone,ethanol and benzene. |
| Formula Weight | 158.17 (Anhydrous) |
| Percent Purity | 99.9965% |
| Grade | Puratronic |
| Sensitivity | Hygroscopic |
| Show More |
RUO – Research Use Only
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.