missing translation for 'onlineSavingsMsg'
Learn More

Calcium acetate hydrate, Puratronic™, 99.9965% (metals basis)

Product Code. 11352068
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11352068 25 g 25g
11362068 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352068 Supplier Thermo Scientific Alfa Aesar Supplier No. 012974.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 114460-21-8 | C4H6CaO4 | 158.17 g/mol

Calcium acetate has been utilized in the crystallization of proteins, such as human annexin II and the bacterial heme oxygenase (Hmu O), and of a complex of human CDK6 with a virus-encoded cyclin. It has been used in the manufactureof metallic soaps and synthetic resins, in lubricants as a corrosion inhibitor, and as a mordant in dyeing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114460-21-8
Molecular Formula C4H6CaO4
Molecular Weight (g/mol) 158.17
MDL Number MFCD00012448
InChI Key VSGNNIFQASZAOI-UHFFFAOYSA-L
IUPAC Name calcium diacetate
SMILES [Ca++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point (decomposition)
Physical Form Powder
Quantity 25 g
Beilstein 3692527
Merck Index 14,1646
Solubility Information Soluble in water. Slightly soluble in methanol,hydrazine insoluble in acetone,ethanol and benzene.
Formula Weight 158.17 (Anhydrous)
Percent Purity 99.9965%
Grade Puratronic
Sensitivity Hygroscopic
Density 1.5 g/mL
Assay (metals basis)
Chemical Name or Material Calcium acetate hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.