missing translation for 'onlineSavingsMsg'
Learn More

Calcium acetate hydrate, 99%, extra pure

Product Code. p-4509929 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
1kg
250g
5kg
This item is not returnable. View return policy

Product Code. 10772251

Brand: Thermo Scientific Acros 211052500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 114460-21-8 | C4H6CaO4 | 158.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114460-21-8
Molecular Formula C4H6CaO4
Molecular Weight (g/mol) 158.17
MDL Number MFCD00012448
InChI Key VSGNNIFQASZAOI-UHFFFAOYSA-L
Synonym Acetic acid, calcium salt hydrate
IUPAC Name calcium diacetate
SMILES [Ca++].CC([O-])=O.CC([O-])=O

Specifications

Color White
pH 7.0 to 9.0 (10% soln., 20°C)
Physical Form Powder and/or Agglomerates
Quantity 250 g
Assay Percent Range 98.5 to 101.5% (on anhydrous substance) (Complexometry)
Linear Formula (CH3CO2)2Ca
Beilstein 02,IV,113
Fieser 13,60; 15,46
Merck Index 14,1646
Solubility Information Solubility in water: 250g/L (20°C). Other solubilities: slightly soluble in alcohol
Formula Weight 158.17
Percent Purity 99%
Grade Extra Pure
Infrared Spectrum Authentic
Packaging Plastic bottle
Chemical Name or Material Calcium acetate hydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.