missing translation for 'onlineSavingsMsg'
Learn More

Calcium 3-hydroxy-3-methylbutyrate hydrate, 97+%

Product Code. 11345487
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11345487 50 g 50g
11325487 250 g 250g
11335487 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11345487 Supplier Thermo Scientific Alfa Aesar Supplier No. 042721.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 135236-72-5 | C10H18CaO6 | 274.33 g/mol

Calcium 3-hydroxy-3-methylbutyrate hydrate is a nutritional supplement, which represents a calcium salt of beta-hydroxy beta-methylbutyrate (HMB). The latter is a leucine metabolite produced in the body through oxidation of the ketoacid of leucine (ketoisocaproate, KIC). Because leucine plays an important regulatory role in protein metabolism and due to the conversion of leucine to KIC, several researchers have presumed that KIC is the active ingredient responsible for most of the beneficial effects of leucine on protein metabolism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135236-72-5
Molecular Formula C10H18CaO6
Molecular Weight (g/mol) 274.33
MDL Number MFCD01318562
InChI Key WLJUMPWVUPNXMF-UHFFFAOYSA-L
Synonym calcium 3-hydroxy-3-methyl butyrate
PubChem CID 131847069
SMILES [Ca++].CC(C)(O)CC([O-])=O.CC(C)(O)CC([O-])=O

Specifications

Color White
Quantity 50 g
Solubility Information Soluble in water.
IUPAC Name calcium bis(3-hydroxy-3-methylbutanoate)
Formula Weight 274.33 (Anhydrous)
Percent Purity ≥97%
Physical Form Powder
Chemical Name or Material Calcium 3-hydroxy-3-methylbutyrate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.