missing translation for 'onlineSavingsMsg'
Learn More

Calcium 2-ethylhexanoate, Superconductor Grade, in 2-ethylhexanoic acid (3-8% Ca)

Product Code. 11425190
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11425190 5 g 5g
11328088 25 g 25g
11338088 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11425190 Supplier Thermo Scientific Alfa Aesar Supplier No. 036657.06
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 136-51-6 | C16H30CaO4 | 326.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136-51-6
Molecular Formula C16H30CaO4
Molecular Weight (g/mol) 326.49
MDL Number MFCD00014001
InChI Key LTPCXXMGKDQPAO-UHFFFAOYSA-L
Synonym calcium 2-ethylhexanoate, calcium bis 2-ethylhexanoate, hexanoic acid, 2-ethyl-, calcium salt, hexanoic acid, 2-ethyl-, calcium salt 2:1, calcium 2-ethyl-hexanoate, octope ca, calcium hex-cem, nikka octhix calcium, calcium 2-ethylhexoate, dsstox_cid_7063
PubChem CID 61082
IUPAC Name calcium;2-ethylhexanoate
SMILES CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Ca+2]

Specifications

Odor Weak, characteristic
Quantity 5 g
Formula Weight 326.5
Grade Superconductor
Concentration or Composition (by Analyte or Components) 3 to 8% Ca
Physical Form Liquid
Chemical Name or Material Calcium 2-ethylhexanoate, in 2-ethylhexanoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.