missing translation for 'onlineSavingsMsg'
Learn More

Calcium 2,4-pentanedionate hydrate, 99%

Product Code. 11327298
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11327298 10 g 10g
11317298 50 g 50g
11307298 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11327298 Supplier Thermo Scientific Alfa Aesar Supplier No. 030526.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 345909-31-1 | C10H14CaO4 | 238.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 345909-31-1
Molecular Formula C10H14CaO4
Molecular Weight (g/mol) 238.30
MDL Number MFCD00013486 MFCD00151667
InChI Key QAZYYQMPRQKMAC-FDGPNNRMSA-L
Synonym calcium bis 4-oxopent-2-en-2-olate hydrate
PubChem CID 16219078
SMILES [Ca++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 175°C (decomposition)
Quantity 10 g
IUPAC Name calcium bis((2Z)-4-oxopent-2-en-2-olate)
Formula Weight 238.30 (Anhydrous)
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material Calcium 2,4-pentanedionate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.