missing translation for 'onlineSavingsMsg'
Learn More

Calcitroic Acid, TRC

Product Code. 30138716
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138716 10 mg 10mg
30134752 5 mg 5mg
30012086 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138716 Supplier Toronto Research Chemicals Supplier No. TRCC14455010MG
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 71204-89-2
Molecular Formula C23 H34 O4
Molecular Weight (g/mol) 374.51
Synonym 1H-Indene-1-propanoic acid, 4-[(2Z)-2-[(3S,5R)-3,5-dihydroxy-2-methylenecyclohexylidene]ethylidene]octahydro-β,7a-dimethyl-, (βR,1R,3aS,4E,7aR)-, 24-Nor-9,10-secochola-5,7,10(19)-trien-23-oic acid, 1,3-dihydroxy-, (1α,3β,5Z,7E)- (9CI), (βR,1R,3aS,4E,7aR)-4-[(2Z)-2-[(3S,5R)-3,5-Dihydroxy-2-methylenecyclohexylidene]ethylidene]octahydro-β,7a-dimethyl-1H-indene-1-propanoic acid, 1α-Hydroxycalcioic acid, Calcitroic acid, MC 1235
IUPAC Name (3R)-3-[(1R,3aS,4E,7aR)-4-[(2Z)-2-[(3S,5R)-3,5-dihydroxy-2-methylidenecyclohexylidene]ethylidene]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-1-yl]butanoic acid
SMILES C[C@H](CC(=O)O)[C@H]1CC[C@H]2\C(=C\C=C/3\C[C@@H](O)C[C@H](O)C3=C)\CCC[C@]12C

Specifikationer

Quantity 10 mg
InChI Formula InChI=1S/C23H34O4/c1-14(11-22(26)27)19-8-9-20-16(5-4-10-23(19,20)3)6-7-17-12-18(24)13-21(25)15(17)2/h6-7,14,18-21,24-25H,2,4-5,8-13H2,1,3H3,(H,26,27)/b16-6+,17-7-/t14-,18-,19-,20+,21+,23-/m1/s1
Formula Weight 374.2457
Chemical Name or Material Calcitroic Acid
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.