missing translation for 'onlineSavingsMsg'
Learn More

SRT3190, MedChemExpress

Product Code. 16475488
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16475488

Brand: Medchem Express HY13021

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SRT3190 is an antagonist of CXCR2, used in the research of chemokine mediated diseases.

SRT3190 is an antagonist of CXCR2, used in the research of chemokine mediated diseases.

Specifications

Chemical Name or Material SRT3190
CAS 1204707-73-2
Color White
Purity Grade Notes Research
Molecular Formula C18H23F2N5O4S2
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (210.29 mM)
SMILES OC[C@@H]([C@@H](NC1=CC(NS(N2CCC2)(=O)=O)=NC(SCC3=C(C(F)=CC=C3)F)=N1)C)O
Molecular Weight (g/mol) 475.53
Formula Weight 475.53
Percent Purity 99.32%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.