missing translation for 'onlineSavingsMsg'
Learn More

Calcitriol Impurities D, MedChemExpress

Product Code. 16428228
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
This item is not returnable. View return policy

Product Code. 16428228

Brand: Medchem Express HY77274

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Calcitriol Impurities D is the impurity of Calcitriol, Calcitriol is the hormonally active form of vitamin D, Calcitriol is the active metabolite of vitamin D3 that activates the vitamin D receptor (VDR).

Calcitriol Impurities D is the impurity of Calcitriol, Calcitriol is the hormonally active form of vitamin D, Calcitriol is the active metabolite of vitamin D3 that activates the vitamin D receptor (VDR).

Specifications

Chemical Name or Material Calcitriol Impurities D
CAS 103656-40-2
Color White
Purity Grade Notes Research
Molecular Formula C28H46O3
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (232.20 mM; Need ultrasonic)
SMILES O[C@H](C/1=C)C[C@H](O)CC1=C\C=C2[C@@](CC[C@]3([H])[C@@H](CCCCC(C)(O)C)C)([H])[C@]3(C)CCC\2
Molecular Weight (g/mol) 430.66
Formula Weight 430.66
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.