missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Calcitonin, Salmon

Product Code. 15455739
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15455739 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15455739 Supplier Thermo Scientific Chemicals Supplier No. J63531.!D
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

It shows hypocalcaemic and hypophosphatemic action, with amino-acid structure differing amongst mammalian species. Salmon calcitonin (salcatonin is the synthetic analogue) shows a markedly different sequence to that of the higher vertebrates as well as more potent activity. It decreases blood calcium and phosphate due to inhibition of resorption by osteoblasts and osteocytes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Species Salmon
Purification Method HPLC
Format Powder
For Use With (Application) A peptide hormone
Molecular Weight (g/mol) 3417.90
Product Type Calcitonin, salmon
Quantity 0.5 mg
Regulatory Status RUO - research use only

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.