missing translation for 'onlineSavingsMsg'
Learn More

Calcitonin, Salmon, TRC

Product Code. 30330091
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
This item is not returnable. View return policy

Product Code. 30330091

Brand: Toronto Research Chemicals TRCC1445582.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Calcitonin (salmon)
CAS 47931-85-1
Molecular Formula C145 H240 N44 O48 S2
Quantity 2.5 mg
Synonym Calcitonin (salmon), 1,2-Dithia-5,8,11,14,17,20-hexaazacyclotricosane, cyclic peptide deriv., Calciben, Calcimar, Calcitonin (salmon reduced) cyclic (1-7)-disulfide, Calcitonin (salmon reduced), cyclic (1-7)-disulfide, Calsyn, Calsynar, Catonin, Karil, L-Prolinamide, L-cysteinyl-L-seryl-L-asparaginyl-L-leucyl-L-seryl-L-threonyl-L-cysteinyl-L-valyl-L-leucylglycyl-L-lysyl-L-leucyl-L-seryl-L-glutaminyl-L-α-glutamyl-L-leucyl-L-histidyl-L-lysyl-L-leucyl-L-glutaminyl-L-threonyl-L-tyrosyl-L-prolyl-L-arginyl-L-threonyl-L-asparaginyl-L-threonylglycyl-L-serylglycyl-L-threonyl-, cyclic (1-7)-disulfide, Miacalcic, Miacalcin, Miadenil, Prontocalcin, Rulicalcin, SMC 021 A/C, Salcatonin, Salmon Thyrocalcitonin, Salmon calcitonin, Salmon calcitonin I, Salmon calcitonin-(1-32), Salmotonin, Stalcin, Tonocalcin, Calcitonin Salmon, Calcitonin salmon
InChI Formula InChI=1S/C145H240N44O48S2/c1-65(2)45-86(175-139(232)110(70(11)12)183-136(229)99-63-239-238-62-79(148)117(210)178-96(59-191)134(227)174-92(52-104(151)202)131(224)172-90(49-69(9)10)129(222)180-98(61-193)135(228)187-114(74(16)197)142(235)181-99)118(211)158-55-106(204)162-80(25-18-20-40-146)120(213)169-89(48-68(7)8)128(221)179-97(60-192)133(226)167-83(34-37-102(149)200)122(215)165-85(36-39-109(207)208)123(216)171-88(47-67(5)6)127(220)173-91(51-77-54-156-64-161-77)130(223)164-81(26-19-21-41-147)121(214)170-87(46-66(3)4)126(219)166-84(35-38-103(150)201)125(218)186-113(73(15)196)141(234)177-94(50-76-30-32-78(199)33-31-76)143(236)189-44-24-29-101(189)137(230)168-82(27-22-42-157-145(154)155)124(217)185-112(72(14)195)140(233)176-93(53-105(152)203)132(225)184-111(71(13)194)138(231)160-56-107(205)163-95(58-190)119(212)159-57-108(206)182-115(75(17)198)144(237)188-43-23-28-100(188)116(153)209/h30-33,54,64-75,79-101,110-115,190-199H,18-29,34-53,55-63,146-148H2,1-17H3,(H2,149,200)(H2,150,201)(H2,151,202)(H2,152,203)(H2,153,209)(H,156,161)(H,158,211)(H,159,212)(H,160,231)(H,162,204)(H,163,205)(H,164,223)(H,165,215)(H,166,219)(H,167,226)(H,168,230)(H,169,213)(H,170,214)(H,171,216)(H,172,224)(H,173,220)(H,174,227)(H,175,232)(H,176,233)(H,177,234)(H,178,210)(H,179,221)(H,180,222)(H,181,235)(H,182,206)(H,183,229)(H,184,225)(H,185,217)(H,186,218)(H,187,228)(H,207,208)(H4,154,155,157)/t71-,72-,73-,74-,75-,79+,80+,81+,82+,83+,84+,85+,86+,87+,88+,89+,90+,91+,92+,93+,94+,95+,96+,97+,98+,99+,100+,101+,110+,111+,112+,113+,114+,115+/m1/s1
SMILES CC(C)C[C@H](NC(=O)[C@@H](NC(=O)[C@@H]1CSSC[C@H](N)C(=O)N[C@@H](CO)C(=O)N[C@@H](CC(=O)N)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CO)C(=O)N[C@@H]([C@@H](C)O)C(=O)N1)C(C)C)C(=O)NCC(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CO)C(=O)N[C@@H](CCC(=O)N)C(=O)N[C@@H](CCC(=O)O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](Cc2c[nH]cn2)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCC(=O)N)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](Cc3ccc(O)cc3)C(=O)N4CCC[C@H]4C(=O)N[C@@H](CCCNC(=N)N)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](CC(=O)N)C(=O)N[C@@H]([C@@H](C)O)C(=O)NCC(=O)N[C@@H](CO)C(=O)NCC(=O)N[C@@H]([C@@H](C)O)C(=O)N5CCC[C@H]5C(=O)N
IUPAC Name (4S)-4-[[(2S)-5-amino-2-[[(2S)-2-[[(2S)-2-[[(2S)-6-amino-2-[[2-[[(2S)-2-[[(2S)-2-[[(4R,7S,10S,13S,16S,19S,22R)-22-amino-16-(2-amino-2-oxoethyl)-7-[(1R)-1-hydroxyethyl]-10,19-bis(hydroxymethyl)-13-(2-methylpropyl)-6,9,12,15,18,21-hexaoxo-1,2-dithia-5,8,11,14,17,20-hexazacyclotricosane-4-carbonyl]amino]-3-methylbutanoyl]amino]-4-methylpentanoyl]amino]acetyl]amino]hexanoyl]amino]-4-methylpentanoyl]amino]-3-hydroxypropanoyl]amino]-5-oxopentanoyl]amino]-5-[[(2S)-1-[[(2S)-1-[[(2S)-6-amino-1-[[(2S)-1-[[(2S)-5-amino-1-[[(2S,3R)-1-[[(2S)-1-[(2S)-2-[[(2S)-1-[[(2S,3R)-1-[[(2S)-4-amino-1-[[(2S,3R)-1-[[2-[[(2S)-1-[[2-[[(2S,3R)-1-[(2S)-2-carbamoylpyrrolidin-1-yl]-3-hydroxy-1-oxobutan-2-yl]amino]-2-oxoethyl]amino]-3-hydroxy-1-oxopropan-2-yl]amino]-2-oxoethyl]amino]-3-hydroxy-1-oxobutan-2-yl]amino]-1,4-dioxobutan-2-yl]amino]-3-hydroxy-1-oxobutan-2-yl]amino]-5-carbamimidamido-1-oxopentan-2-yl]carbamoyl]pyrrolidin-1-yl]-3-(4-hydroxyphenyl)-1-oxopropan-2-yl]amino]-3-hydroxy-1-oxobutan-2-yl]amino]-1,5-dioxopentan-2-yl]amino]-4-methyl-1-oxopentan-2-yl]amino]-1-oxohexan-2-yl]amino]-3-(1H-imidazol-4-yl)-1-oxopropan-2-yl]amino]-4-methyl-1-oxopentan-2-yl]amino]-5-oxopentanoic acid
Molecular Weight (g/mol) 3431.85
Formula Weight 3429.7133
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.